Platinum ≥99.99% (metals basis), foil, Premion®, Thickness: 0.25 mm (0.01 in)

Supplier: ALFA AESAR

11508.FF 11508.FI 11508.GH
11508.FFEA 884 EUR
11508.FF 11508.FI 11508.GH
Platinum ≥99.99% (metals basis), foil, Premion®, Thickness: 0.25 mm (0.01 in)
Platinum

Purity: 99.99% CAS No.: 7440-06-4 Physical Form: ≈3.35g/25x25mm Odour: Odourless MDL No.: MFCD00011179

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR