Palladium ≥99.9% (metals basis), foil, Thickness: 0.25 mm

Supplier: ALFA AESAR

11516.FF 11516.FI 11516.GH
11516.FFEA 725 EUR
11516.FF 11516.FI 11516.GH
Palladium ≥99.9% (metals basis), foil, Thickness: 0.25 mm
Palladium
Formula: Pd
MW: 106.420 g/mol
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR