Ammonium sulphate ≥99.0% ACS

Supplier: ALFA AESAR
Warning

11566.22 11566.36 11566.A3 11566.A9
11566.22EA 25.3 EUR
11566.22 11566.36 11566.A3 11566.A9
Ammonium sulphate ≥99.0% ACS
Ammonium sulphate

Purity: 99.0% min CAS No.: 7783-20-2 Molecular Formula: H8N2O4S Formula Weight: 132.14 Physical Form: Granular Density: 1.769 Appearance: White Melting Point: ca 280° dec. Solubility: Soluble in water. Insoluble in alcohol and acetone Merck Reference: 14,555 MDL No.: MFCD00003391


In galvanizing steel, manufacturing of ammonia alum and sulfuric acid

Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: ∼ 280 °C (dec.)
Density: 1.769
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 14,555

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR