Lysing matrix Z, MP Biomedicals
Supplier: MP BIOMEDICALS
116961100 116540438 116961001 116961050 116961500 116978025 116978050 116985050 116985025 116985100 116978005 116540439
ICNA116961100EA 357 EUR
ICNA116961100 ICNA116540438 ICNA116961001 ICNA116961050 ICNA116961500 ICNA116978025 ICNA116978050 116985050. 116985025. 116985100. 116978005. 116540439.
Lysing matrix Z, MP Biomedicals
Protein Extraction Protein Extraction Reagents
Lysing Matrix Z is used primarily for lysis of animal and plant tissue, tougher stems, roots and seeds, and whole insects. Each impact-resistant tube contains yttria-stabilised zirconium oxide beads.
  • Impact-resistant tube
  • Ideal for cracking of very tough cell walls
  • Lower shear rate minimises degradation
  • Free from DNase and RNase
Matrix Z is an alternative to the very aggressive Matrix A because it's lower shear rate minimises degradation in applications such as RNA and protein isolation from very tough tissue.
Order Now
SPECIFICATIONS
Environmentally Preferable
Sample type Plants; Animal Tissues; Wood; Seeds; Soil; Clay; Bacteria; Insects; Ancient and Dried Samples