Rhodium ≥99.95% (metals basis), sponge -20 mesh

Supplier: ALFA AESAR
Warning

12073.04
12073.04EA 4710 EUR
12073.04
Rhodium ≥99.95% (metals basis), sponge -20 mesh
Rhodium

Purity: 99.95% CAS No.: 7440-16-6 Odour: Odourless UN No.: 3089 MDL No.: MFCD00011201

Formula: Rh
MW: 102.9 g/mol
Melting Pt: 1960
Density: 12.41
Storage Temperature: Ambient
MDL Number: MFCD00011201
CAS Number: 7440-16-6
EINECS: 231-125-0
UN: 3089
ADR: 4.1 III
Merck Index: 12,08353

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR