Sodium cyanide ≥95% ACS

Supplier: ALFA AESAR
Danger

12137.36 12137.A3 12137.22
12137.36EA 101 EUR
12137.36 12137.A3 12137.22
Sodium cyanide ≥95% ACS
Sodium cyanide

Purity: 95% min CAS No.: 143-33-9 Molecular Formula: NaCN Formula Weight: 49.01 Physical Form: Granular Density: 1.61 Melting Point: 563° Boiling Point: ca 1500° Solubility: Soluble in water Beilstein Reference: 3587243 Merck Reference: 14,8605 UN No.: 1689 MDL No.: MFCD00003523

Formula: CNNa
Boiling Pt: ∼1500 °C
Melting Pt: 563 °C
Density: 1,61
Storage Temperature: Ambient
MDL Number: MFCD00003523
CAS Number: 143-33-9
EINECS: 205-599-4
UN: 1689
ADR: 6.1,I
Merck Index: 14,8605

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR