Aluminium oxide 20% in water, colloidal dispersion

Supplier: ALFA AESAR

12733.A1 12733.4Z
12733.A1EA 54.8 EUR
12733.A1 12733.4Z
Aluminium oxide 20% in water, colloidal dispersion
Aluminium oxide

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Physical Form: 0.05 Micron Particles Odour: Odourless MDL No.: MFCD00003424


Applications: Effective in high temperature refractories as a stable binder

Formula: Al₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR