VWR®, Biohazard Bags, PP
Supplier: VWR Collection
129-0035EA 580 EUR
129-0035 129-0037 129-0033 129-0034 129-1550 129-1551 129-1552 129-1553 129-1556 129-1557 129-1558 129-1559 129-1554
VWR®, Biohazard Bags, PP
Bags Waste Disposal Bags
These sturdy bags are ideal for disposing of biologically hazardous waste that must first be autoclaved. PP, 50 µm thick.
  • Autoclavable up to 135 °C
  • Available with or without temperature indicator
  • Useful temperature indicator text is white before autoclaving, then black
  • Text 'Contents Have Been Autoclaved' indicates successful autoclaving
  • Luminous red for easy recognition and highly visible 'Biohazard' label with corresponding warning sign
Bags are formulated to withstand stress at the side seams and are strong enough to hold items in the laboratory.
More Product Information