VWR®, Flat Cleanroom Mops
Supplier: VWR Collection
129-0067EA 506 EUR
129-0067 129-0068 129-0069 129-0070 129-0071 129-0072
VWR®, Flat Cleanroom Mops
Mops
Foam with 100% polyester cover. For applying disinfectants during pharmaceutical production or for aseptic processes. The flat mop head is ideally suited for cleaning large surfaces such as floors, walls and ceilings. Low profile, thick padding and angled head for ease of use.
  • Autoclavable, washable and reusable
  • Good moisture bonding provides optimum contact with the disinfectant
  • Available in sterile and non sterile versions
  • Resistant to strong disinfectants
Suitable for Class 10, ISO 4, M2 cleanrooms.
Ordering information: The complete mopMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable