Buckets, TruCLEAN™
Supplier: PERFEX
22-15R 22-15W 22-06B 22-06R 22-06W 22-15B
129-0106EA 153 EUR
129-0106 129-0107 129-0111 129-0113 129-0112 129-0108
Buckets, TruCLEAN™
Buckets
Temperature resistant polypropylene buckets with stainless steel handles.
  • Buckets interlock when placed side by side
  • Non-porous, super smooth surface prevents microbial growth
  • Embossed graduations in English for accurate measure
  • Stainless steel handles will not rust, corrode or oxidize
  • Autoclavable, gamma ETO sterilization compatible
  • Impervious to most chemicals, solvents and detergents
Buckets are seated firmly within cart base eliminating any possibility of spillage during transport.
Ideal for any critical manufacturing environment. CompatibleMore Product Information