Wall washing systems, VertiKlean®
Supplier: CONTEC
LVK-MOP MVK-MOP VK-MOPIR LVK-MOPIR MVK-MOPIR TLVKMOP LVK-SOX MVK-QT-SOX VK-MOP 2652K 2652SS TMVKMOP TLVKMOPIR 2653SS 2652LF 2651LF
129-0157EA 103 EUR
129-0157 129-0158 141-0988 141-0971 141-0972 141-0982 129-0977 129-0978 141-0987 141-1468 141-1465 141-1660 141-1661 141-1466 141-1453 CONT2651LF
Wall washing systems, VertiKlean®
Mops
These lightweight, polyester disposable mops are excellent for applying disinfectants and for wet cleaning of vertical surfaces, ceilings, and walls in controlled environments. Ideal for reaching awkward corners and ceilings with a complete range of motion.
  • Mop heads contain no cellulose and are compatible with a wide range of chemicals, detergents, disinfectants and cleaners
  • Mop heads are laser cut to seal the edges, further limiting potential contamination in the mopping process
  • Heads are intended for one-time use
  • Mop heads are available non sterile and
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable