Cable ties, non-releasable, stainless steel
Supplier: AZPACK
129-1134 129-1136
129-1134EA 398 EUR
129-1134 129-1136
Cable ties, non-releasable, stainless steel
Ties
These stainless steel ties are designed to secure hoses, cables, poles, pipes, and more when harsh environmental conditions may adversely affect the bundling application.
  • Quick and easy installation, either by hand or by tensioning tool
  • Fully enclosed head does not allow dirt or grit to interfere with locking mechanism
  • Marine grade uncoated
  • Operating temperature from −80 to +538 °C
Used where corrosion, vibration, weathering, radiation, and temperature extremes are a concern. The ties can be used in virtually any indoor, outdoor, and undergroundMore Product Information