VWR® MEDBAG, Hospital and Laboratory Sampling Bags
Supplier: VWR Collection
129-1175EA 384 EUR
129-1175 129-1176
VWR® MEDBAG, Hospital and Laboratory Sampling Bags
Bags Sample Bags
VWR® MEDBAGs are liquid-tight bags, used as internal hospital and external secondary packaging for the transportation of category B biological substances (UN 3373). They are the ideal solution for biological and medical transportation. Designed for blood sampling sectors, they combine the highest levels of leak resistance and tracking technologies to bring real benefits to multiple analytical processes.
  • Easy to use, it guarantees a perfect seal with a fast adhesive closure and has quick and easy opening
  • Its unique opening system gives perfect leak
More Product Information