Sterlisation gusseted pouches, Stericlin®
Supplier: Vereinigte Papierwarenfabriken GmbH
3FKSB220108 3FKSB220116 3FKSB220112 3FKSB220118 3FKSB220114 3FKSB220102 3FKSB220104 3FKSB220106 3FKSB220110
129-1266EA 481 EUR
129-1266 129-1270 129-1268 129-1271 129-1269 129-1263 129-1264 129-1265 129-1267
Sterlisation gusseted pouches, Stericlin®
Bags Sterilisation Pouches
These see-through sterilisation pouches made of sterilisation proof 70 g/m² heavy-duty paper are a universal packaging solution.
  • Effective barrier against germs and bacteria
  • Permits the transmission of air, steam or sterilisation gases
  • Three indicators for STEAM-, EO- and FORM-sterilisation are imprinted outside the filling space
Suitable for sterilisation applications of almost all light and medium weight instruments and sets.
Certifications: DIN EN ISO 11607 and DIN 868-5.
Order Now
SPECIFICATIONS
Environmentally Preferable