Thermostatically controlled cabinets, TC series
Supplier: Lovibond Tintometer
2438200-V 2438210-V 2438270-V 2438250-V 2438260-V
132-0954EA 2250 EUR
132-0954 132-0955 LOVI2438270-V LOVI2438250-V LOVI2438260-V
Thermostatically controlled cabinets, TC series
Incubators BOD Incubators
The TC series of thermostatically controlled cabinets is used for continuous temperature control over a range of 2 to 40 °C. This makes them ideal for a wide range of different applications in industrial and research laboratories. In particular they are ideal for the temperature controlled storage of samples or BOD determination in effluent analysis work. Devices such as magnetic stirrers, which require a power supply, can be connected to sockets incorporated in the interior of the cabinet. Improved, robust, insulated housing and highly efficient components provide maximum energy efficiency.
More Product Information