Vertical laminar flow cabinets, DLF series
Supplier: CleanAir by Baker
NP0024360 NP0037460 NP0024460 NP0024560 NP0024660 NP0037360
135-0078EA 10640 EUR
135-0078 CLEANP0037460 CLEANP0024460 CLEANP0024560 CLEANP0024660 CLEANP0037360
Vertical laminar flow cabinets, DLF series
Fume Hoods and Enclosures Laminar Flow Cabinets
CleanAir by Baker DLF series is a high quality vertical laminar flow workbench range for handling non-hazardous materials that require a clean working environment. DLF PCR Series is designed to perform PCR. These cabinets ambient air entering the work area, ensures product protection.
  • Microprocessor flow control
  • Smooth interior for efficient and easy cleaning
  • Control control system with alarms and parameters for work mode, UV light, gas valve
  • Epoxy coated steel interior and exterior (1,5 to 2 mm)
  • Stainless Steel worktop
  • Sliding window (safety glass)
More Product Information