Heraguard™ ECO Horizontal Laminar Flow Cabinets
Supplier: Heraeus
51029702 51029703 51029704 51029701
135-1738EA 8330 EUR
135-1738 135-1739 135-1740 135-1737
Heraguard™ ECO Horizontal Laminar Flow Cabinets
Fume Hoods and Enclosures Laminar Flow Cabinets
Heraguard™ ECO Clean Bench cabinets are ideal for laboratory applications where sample protection is required. Due to DC motors, Heraguard™ ECO Clean Bench cabinets use less energy to operate so there is less heat emitted into the laboratory.
  • Easy to use, intuitive control panel
  • All-round vision: The side windows increase the light inside the work area and support a comfortable work environment
  • Easy pre-filter and HEPA filter exchange
  • Transparent hinged windscreen protects the user
  • Side panels made from single pane safety glass, with an access port for
More Product Information