Production and laboratory chairs, with permanent contact backrest, Neon 3
Supplier: bimos
2000-3278-525 2000-3279-525 2000-3280-525 2001-3278-525 2001-3279-525 2001-3280-525 MG01-3278-525 MG01-3279-525 MG01-3280-525 MG013279525807 20003278525807 MG013278525807 20013280525807 20013279525807 20013278525807 20003280525807 20003279525807 MG013280525807 2002-3278-525 2002-3278-807 2002-3279-525 2002-3279-807 2002-3280-525 2002-3280-807 95616802328058 95616801327958 95616801328052 95616801328058 95616802327852 95616802327858 95616802327952 95616802328052 95616801327852 95616811327852 95616811327858 95616811327952 95616811327958 95616811328052 95616811328058 95616802327958 95616801327952 95616801327858 9561023279807 9561-2000-3277 9561-6801-3277 9561-MG01-3277 9561-2000-3278 9561-MG01-3278 9561-MG02-3280 9561-2000-3279 95612000327852 9561MG01327752 9561-MG01-3279 95616907327852 9561MG013277 95612000327980 9561MG01327980 9561E-MG01-327 9561-SP02-3278 9561-SP02-3277 9561-SP02-3279 9561-SP02-3280 9561-SP01-3280
132-1430EA 688 EUR
132-1430 132-1431 132-1432 132-1433 132-1434 132-1435 132-1448 132-1449 132-1450 132-0905 132-0901 132-0904 BIMO20013280525807 BIMO20013279525807 132-1436 132-0903 132-0902 132-0906 132-1437 132-1438 132-1439 132-1440 132-1441 132-1442 BIMO95616802328058 132-0908 BIMO95616801328052 132-0909 BIMO95616802327852 BIMO95616802327858 BIMO95616802327952 BIMO95616802328052 BIMO95616801327852 BIMO95616811327852 BIMO95616811327858 BIMO95616811327952 BIMO95616811327958 BIMO95616811328052 BIMO95616811328058 BIMO95616802327958 BIMO95616801327952 132-0907 BIMO9561023279807 132-0976 132-0978 132-0977 139-0629 139-0630 BIMO9561-MG02-3280 BIMO9561-2000-3279 BIMO95612000327852 BIMO9561MG01327752 BIMO9561-MG01-3279 BIMO95616907327852 BIMO9561MG013277 BIMO95612000327980 BIMO9561MG01327980 BIMO9561E-MG01-327 BIMO9561-SP02-3278 BIMO9561-SP02-3277 BIMO9561-SP02-3279 BIMO9561-SP02-3280 BIMO9561-SP01-3280
Production and laboratory chairs, with permanent contact backrest, Neon 3
Chairs
Neon supports the forward-leaning sitting position that is often necessary in lab work. Its modern design matches the high-tech laboratory environment perfectly and of course it has all the characteristics you would expect of a good laboratory chair. It is seamless, easy to clean, washable, and resistant to disinfectants. So, Neon is the ideal solution for the laboratory, when a really comfortable ergonomic chair is needed.
  • Permanent contact backrest
  • Ergonomic package with height, seat inclination, seat depth and backrest height adjustment plus weight
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable