Disinfectant, ClearKlens IPA 70%, sterile
Supplier: Diversey
7516050 100865612 7514372
DIVY7516050EA 266 EUR
DIVY7516050 141-0889 DIVY7514372
Disinfectant, ClearKlens IPA 70%, sterile
Cleaning Agents Disinfectants
Disinfectant containing 70% isopropyl alcohol, 30% water for injection, for use on contact surfaces, gloved hands, instruments and utensils.
  • Sterile disinfectant with 70% isopropanol / 30% WFI
  • Ready to use product for the disinfection of work surfaces and process equipment in cleanrooms
  • 0,2 μm filtered, gamma irradiated/aseptically filled and double-bagged
  • ClearKlens® IPA 900 ml trigger spray is an anti-suck back, 'bag in bottle' delivery system to eliminate product contamination during use
  • ClearKlens® IPA 1 L bottle/5 L canister is used for mopping and
More Product Information