Disinfectants, ClearKlens Tego® 2000
Supplier: Diversey
7516051 7514341 7513387
141-0891EA 325 EUR
141-0891 DIVY7514341 141-1220
Disinfectants, ClearKlens Tego® 2000
Cleaning Agents Disinfectants
Highly effective, active amphoteric, surface disinfectant with a broad spectrum of antimicrobial activity, developed for use in sterile environments.
  • Ready to use, 0,2 µm filtered, sterile filled and double-bagged
  • Validated HPLC and titration test methods available
  • Compatible with ClearKlens Cleansinald and ClearKlens Bi-Spore as a rotational process
Certifications: Passes CEN 1276, CEN 1650 and CEN 13697
Caution: Use biocides safely. Always read the label and product information before use.
Articles may not be available in all countries. PleaseMore Product Information