Quantitative filter papers, ashless grades, grade 40, Whatman™
Supplier: GE Healthcare - Whatman
1440-032 1440-042 1440-047 1440-055 1440-070 1440-090 1440-110 1440-125 1440-150 1440-185 1440-240 1440-917 1440-012 1440-320 1440-329 1440-6168 10380004 10380005 10380006
1440032.EA 29.5 EUR
1440032. 1440042. 1440047. 1440055. 1440070. 1440090. 1440110. 1440125. 1440150. 1440185. 1440240. 512-0651 514-0591 514-0592 514-0593 514-0594 WHAT10380004 WHAT10380005 WHAT10380006
Quantitative filter papers, ashless grades, grade 40, Whatman™
Filters Filter Papers Quantitative Filter Paper
Whatman Grade 40 high-purity filters from Cytiva's business are general-purpose papers for quantitative analysis. Grade 40 filters are suitable for a variety of analytical filtration applications.
  • High-purity filters contain 0,007% maximum ash content
  • Medium nominal particle retention rating of 8 µm
  • Medium filtration speed (Herzberg 340 s)
  • Particularly useful for gravimetric analysis
High-purity filters support accurate analysis in applications sensitive to very low levels of background contamination and in gravimetric analysis where ash content mightMore Product Information