Ammonium hydrogen sulphate ≥99.9% (metals basis)

Supplier: ALFA AESAR
Danger

14485.14 14485.22 14485.36
14485.14EA 52.7 EUR
14485.14 14485.22 14485.36
Ammonium hydrogen sulphate ≥99.9% (metals basis)
Ammonium hydrogen sulphate

Purity: 99.9% CAS No.: 7803-63-6 Molecular Formula: NH4HSO4 Formula Weight: 115.10 Physical Form: Lump Sensitivity: Hygroscopic UN No.: 2506 MDL No.: MFCD00051073

Formula: H₅NO₄S
MW: 115.11 g/mol
Boiling Pt: 350
Melting Pt: 146.9 °C
Density: 1.787 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051073
CAS Number: 7803-63-6
EINECS: 232-265-5
UN: 2506
ADR: 8 II
Merck Index: 12,00526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR