Laboratory glass cleaner, deconex® 11 UNIVERSAL
Supplier: Borer Chemie
500100.00-F10W 500100.00-K05W 500100.00-K10W
148-5101EA 38.7 EUR
148-5101 148-5102 148-5103
Laboratory glass cleaner, deconex® 11 UNIVERSAL
Cleaning Agents Detergents and Surface Cleansers
Liquid, mildly alkaline cleaning concentrate for manual cleaning of laboratory glassware and reusable laboratory materials in ultrasonic and soaking baths.
  • Rinsable without residues and guarantees therefore consistent and error-free results
  • With a broad cleaning spectrum
  • Perfect results even at low concentrations
  • Working solution suitable for multiple use
  • Contains active chlorine as a cleaning booster and is designed to replace chromosulfuric acid
  • EDTA- and NTA-free