Imidazole, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS
Danger

1747.0100EA 79.78 EUR
1747.0100 1747.9010
Imidazole, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Imidazole
Formula: C₃H₄N₂
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 89…90 °C
Density: 1,18 g/cm³ (20 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 2923
ADR: 8,III
Merck Index: 13,04935
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=ddb38a8e40ebda2af69627b5ed326af620745f06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay min. 99.5 %
Iron (Fe) max. 0.001 %
Heavy Metals (as Pb) max. 0.001 %
Melting Point 88 - 90 °C
Residue after Ignition max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR