Copper selenide ≥99.5% (metals basis)

Supplier: ALFA AESAR
Danger

17901.06 17901.14
17901.06EA 81 EUR
17901.06 17901.14
Copper selenide ≥99.5% (metals basis)
Copper selenide

Purity: 99.5% CAS No.: 1317-41-5 Molecular Formula: CuSe Formula Weight: 142.50 Physical Form: 6mm & down pieces Density: 6.0-6.6 Melting Point: 550° dec. Solubility: Soluble in HCl with H2Se evolution, in H2SO4 with SO2 evolution. Oxidized to CuSeO3 by HNO3 Merck Reference: 14,2651 UN No.: 3077 MDL No.: MFCD00016058


In semiconductors, as catalyst in Kjeldahl digestion

Formula: CuSe
MW: 142.51 g/mol
Melting Pt: 550 °C (dec.)
Density: 6.0…6.6
Storage Temperature: Ambient
MDL Number: MFCD00016058
CAS Number: 1317-41-5
EINECS: 215-272-8
UN: 3077
ADR: 9 III
Merck Index: 14,2651

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR