Ball valves, VK series
Supplier: VACUUBRAND
20665504 20665505 20665506 20675504 20675505 20675506
181-0034EA 284 EUR
181-0034 VACU20665505 VACU20665506 183-1111 183-1112 183-1113
Ball valves, VK series
Valves Vacuum Valves
Valves of the VK series are sturdy isolation devices for the rough and fine vacuum range. The VKE series uses stainless steel for the metal parts and fiberglass reinforced PTFE seatings.
Ball valves are used for load locks, sample holders, thermocouples or helium supply lines.
Order Now
SPECIFICATIONS
Environmentally Preferable