Tetrabutylammonium bromide 99+%

Supplier: ACROS ORGANICS
Warning

185680250 185681000 185685000 185680500 185680025
185680250.EA 58.6 EUR
185680250. 185681000. 185685000. 185680500. 185680025.
Tetrabutylammonium bromide 99+%
Tetrabutylammonium bromide
Formula: C₁₆H₃₆BrN
Melting Pt: 100…104 °C
Density: 1,148 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011633
CAS Number: 1643-19-2
EINECS: 216-699-2

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥99.0 %
Water ≤1 %
HPLC ≥94 % (sum of tylosins A+B+C+D)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR