Trimethylaluminium 1.0 M in heptane, AcroSeal®

Supplier: ACROS ORGANICS
Danger

189271000 189278000
189271000.EA 98.3 EUR
189271000. 189278000.
Trimethylaluminium 1.0 M in heptane, AcroSeal®
Trimethylaluminium
Formula: C₃H₉Al
MW: 72.09 g/mol
Boiling Pt: 125
Melting Pt: 15
Density: 0.688 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008252
CAS Number: 75-24-1
EINECS: 200-853-0
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration Complexometric 9.5 to 10.5 % w/w (on Al basis)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR