Azur II eosinate, powder, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Danger


M-Clarity™ quality level = MQ100

200220-100G 200220-25G
200220-100GEA 413 EUR
200220-100G 200220-25G
Azur II eosinate, powder, Sigma-Aldrich®
Azur II eosinate
Formula: C₅₁H₄₀Br₄Cl₂N₆Na₂O₅S₂
Storage Temperature: Room Temperature
MDL Number: MFCD00084753
CAS Number: 53092-85-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56dc6044a3f48656fb942fede22b70e7a147538c&vid=56dc6044a3f48656fb942fede22b70e7a147538c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR