Multiple distributors for bottles with GL 45 thread, BOLA
Supplier: Bohlender
D614-08 D615-08
201-1710EA 98.1 EUR
201-1710 201-0299
Multiple distributors for bottles with GL 45 thread, BOLA
Closures Bottle Closures
PPS screw cap, black, distributor body made from PTFE or PFA.
  • Excellent chemical resistance
  • Temperature resistant from −200 to +250 °C
  • Suitable for vacuum applications
For the removal or filling of aggressive or pure liquids. For the connection of hoses, tubes and probes into vessels. Hoses with a maximum external diameter of  8,5 mm can be inserted through the necks.