Jacketed beakers with hose connections
Supplier: NEUBERT VOLUME GLASSWAERE
0710-0250-55 0710-2000-112
201-0372EA 383 EUR
201-0372 201-0374
Jacketed beakers with hose connections
Beakers
Borosilicate glass 3.3, without spout.
  • Very good chemical resistance
  • Very good thermal resistance
  • With glass tubing connectors O.Ø: 10 mm
For maintaining liquid samples at a constant temperature. The temperature controlling medium is pumped through the double-walled jacket by means of a recirculating thermostat.