VWR® Expansion Adapters for Conical Ground Joints
Supplier: VWR Collection
201-1447EA 16.8 EUR
201-1447 201-1448
VWR® Expansion Adapters for Conical Ground Joints
Adapters Adapters, Conical Ground Joints
Adapters from small cone at the top to large socket at the bottom.
  • Made from clear borosilicate 3.3 glass
  • Excellent chemical resistance, high temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • Outstanding leak tightness
Certifications: Produced according to DIN 12257.