Quinaldine red 95% Dye, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY


MQ200

201316-25G 201316-5G
201316-25GEA 424 EUR
201316-25G 201316-5G
Quinaldine red 95% Dye, Sigma-Aldrich®
Quinaldine red

Quinaldine red has been used as a fluorescent probe for α1-acid glycoprotein (AAG). It is also used a diagnostic assay manufacturing, hematology, histology.


Quinaldine red (2-(p-Dimethylaminostyryl) quinoline ethiodide) is a very dark red or reddish-black colored powder. It has been used mainly for regulation of cell proliferation, in diagnostic agents of disease related with amyloid accumulation, detecting enzyme activity and dental bleaching materials. In addition, quinaldine red also been implicated in treatment of allodynia and virus infectious diseases. It is colorless at pH 1,4 and red at pH 3,2.

Formula: C₂₁H₂₃IN₂
MW: 430.33 g/mol
Melting Pt: 240
Storage Temperature: Room Temperature
MDL Number: MFCD00011968
CAS Number: 117-92-0
EINECS: 204-221-5
Merck Index: 12,08230
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d4684321b9d88857feec4f61191f7654f87da7e&vid=8d4684321b9d88857feec4f61191f7654f87da7e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour) Green to Black
Appearance (Form) Powder to Crystalline Powder
Infrared spectrum Conforms to Structure
Wavelength 279 - 285 nm
Extinction Coefficient ≥8800
Carbon 55.4 - 61.8%
Nitrogen 6.1 - 6.9%
Solubility (Turbidity) Clear
Solubility (Colour) Conforms to requirements dark red to purple


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR