Citric acid, anhydrous 99.5-100.5%, powder Ph. Eur.

Supplier: VWR Chemicals
Warning

20282.293EA 102 EUR
20282.293 20282.362 20282.464
Citric acid, anhydrous 99.5-100.5%, powder Ph. Eur.
Citric acid

Caution: Not suitable for parenteral use

Formula: HOC(COOH)(CH₂COOH)₂
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 149…151 °C
Density: 1,665 g/cm³ (18 °C. OECD 109)
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
REACH: 01-2119457026-42
Merck Index: 13,02350
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=431351cfe967d5cecc0f339b1ca8b269da45dbb8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated on anhydrous) 99.5 - 100.5 %
Appearance Conforms (see CoA/CoS)
Identification B Passes test
Appearance of solution Passes test
Readily carbonisable substances Passes test
Oxalic acid Max. 360 ppm
SO4 (Sulphate) Max. 150 ppm
Heavy metals (as Pb) Max. 10 ppm
Water Max. 1.0 %
Sulphated ash Max. 0.1 %
Residual solvents Unlikely by manuf.process


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR