Isoamyl alcohol, dehydrated (max. 0.05% H₂O) ≥98%, GPR RECTAPUR® for synthesis

Supplier: VWR Chemicals
Warning

20796.298EA 86.1 EUR
20796.298
Isoamyl alcohol, dehydrated (max. 0.05% H₂O) ≥98%, GPR RECTAPUR® for synthesis
Isoamyl alcohol
Formula: (CH₃)₂CHCH₂CH₂OH
MW: 88.15 g/mol
Boiling Pt: 130
Melting Pt: –117 °C
Density: 0.812 g/cm³ (20 °C)
Flash Pt: 42
Storage Temperature: Ambient
MDL Number: MFCD00002934
CAS Number: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3 III
Merck Index: 13,05214
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=60e27777862a1441bdec89c868acf27847ec49b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated as mixture of isomers) Min. 98 %
Density (20/4) 0.808 - 0.812
Water Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR