Test tubes with SVL® screwcaps
Supplier: QVF LABORTECHNIK
611-55
212-1534EA 17.9 EUR
212-1534
Test tubes with SVL® screwcaps
Tubes Test Tubes
PYREX®, borosilicate glass with phenoplastic screw caps.
  • Excellent chemical resistance, excellent thermal resistance
  • Screw cap with PTFE coated seal
  • Caps can be repeatedly dry or steam sterilised (121 °C over 2 hours)
Specially designed for use as culture tubes.