VWR®, Measuring Beakers with Handle
Supplier: VWR Collection
213-3401EA 70.3 EUR
213-3401 213-3402 213-3403 213-3404
VWR®, Measuring Beakers with Handle
Beakers
PP, highly translucent.
  • Good chemical resistance
  • Resistant to heat up to +121 °C (for 20 minutes), prolonged use +100 °C
  • Smooth surfaces for easy, fast cleaning
  • With embossed scale, blue graduations and spout
Ideal for general laboratory use and a wide variety of other applications.
Certifications: Food-safe products acc. to EC Directive No. 10/2011.