VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
215-1592EA 106 EUR
215-1592 215-1593 215-1594 215-0057 215-0058 215-0059 215-1595 215-1596 215-3261
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Bottles Glass Bottles
Borosilicate 3.3 glass, with PP screw cap and pouring ring.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Economical alternative for a wide range of laboratory applications. Round, with graduations, DIN GL 32 or GL 45 thread.
ISO 4796
Certifications: ISO 4796