Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
218011453 218011753 218012458 218013651 218014459 218015455 218017353 218018658 218019157 218018855 218012955 218015155 218016957 218010851 218016357 218019251
215-1512EA 22.4 EUR
215-1512 215-1513 215-1514 215-1515 21801445. 215-1517 215-1519 215-1520 21801915. 215-1521 215-2428 215-2429 215-2430 215-3214 215-1518 215-4775
Laboratory bottles, narrow neck, with screw cap, DURAN®
Bottles Glass Bottles
DURAN®, borosilicate glass 3.3, clear, with PP screw cap.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
Round, with graduation, DIN thread, pouring ring and blue screw cap.
With printed Retrace Code (batch identification).
*With specially shaped glass rim for improved pouring; no additional pouring ring is needed.
**Ring-shaped reinforcement on the bottle shoulder makes the filling line of the nominal volumeMore Product Information