Specimen containers, disposable

Supplier: Simport Scientific
C581-500 C580-3 C580-4 C580-5 C581-1L C581-300 C581-2600 C581-5700
216-1005EA 190 EUR
216-1005 216-1001 216-1002 216-1003 SIMPC581-1L SIMPC581-300 SIMPC581-2600 SIMPC581-5700
Specimen containers, disposable
Sample Containers General Purpose Containers
These containers and closures are made of PP. They are ideal for collection, transport and storage of fluids, powders, solids, pathology specimens and hazardous samples.

  • Disposable
  • Resist temperatures from −20 to + 80 °C

Tight-fitting lids prevent leaks and odors. Certain models listed below have a tight-fitting lid. Also offered with an incorporate tamperproof lid which is opened by first removing tab on side of lid, then by lifting the cover.

The 2600 and 5700 ml sizes are provided with a plastic handle to facilitate handling.

Caution: *TheMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR