Stoppers, rubber, with reversible edge
Supplier: Saint-Gobain
407007-100 407010-50 407015-20 407018-20 407025-10
217-0182EA 83.6 EUR
217-0182 217-0183 391-2258 217-0184 217-0185
Stoppers, rubber, with reversible edge
Stoppers Rubber Stoppers
The white stoppers made of natural rubber guarantee absolute impermeability, even when the edges of the tubes or glass, metal or plastic containers to be sealed are imperfect. They can be punctured and resealed after sampling, even after several uses.
  • Outstanding sealing and protection from the outside atmosphere
  • Suitable for puncturing
  • Economical
Order Now
SPECIFICATIONS
Environmentally Preferable