tert-Butyl methyl ether ≥99.5%, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals
Danger


Filtered through a 0.2 µm filter, packaged under nitrogen.

22105.295EA 116 EUR
22105.295 22105.320
tert-Butyl methyl ether ≥99.5%, HiPerSolv CHROMANORM® for HPLC
tert-Butyl methyl ether
Formula: (CH₃)₃COCH₃
MW: 88.15 g/mol
Boiling Pt: 55.2 °C (1013 hPa)
Melting Pt: –108.6 °C
Density: 0.7405 g/cm³ (20 °C)
Flash Pt: −28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3 II
Merck Index: 13,06059
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=b1aeff56d6e354e402e4eb67a164ec8ddd884ae7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.5 %
Acidity Max. 0.001 meq/g
Evaporation residue Max. 20 ppm
Peroxides Max. 5 ppm
Water Max. 0.05 %
Transmittance (254 nm) Min. 60 %
Transmittance (280 nm) Min. 80 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR