Drum pumps
Supplier: BURKLE
5600-1251 5600-1252 5600-0501 5600-0502 5600-0651 5600-0652 5600-0801 5600-0802 5600-1002 5600-1001 5600-1501 5600-1502
223-1224EA 305 EUR
223-1224 223-1230 223-1220 223-1226 223-1221 223-1227 223-1222 223-1228 223-0077 223-0075 223-0076 223-0078
Drum pumps
Pumps Drum/Transfer Pumps
Drum pumps in PP, with rigid discharge tube or flexible PVC discharge hose with stopcock. Fast simple decanting of liquids from drums, canisters and tanks.
  • Very good chemical resistance, therefore also suitable for decanting acids, alkalis and aggressive cleaning agents
  • Delivery rate up to approximately 12 L/min
  • Stepless adjustment of pump immersion depth, ensuring good residual extraction
  • Flexible PVC discharge hose is 1,2 m long
  • The low level base non return valve ensures good removal of the liquid remaining in the barrel
Order Now
SPECIFICATIONS
Environmentally Preferable