Bench protectors, Benchkote® Plus, Whatman™
Supplier: GE Healthcare - Whatman
2301-6160
23016160.EA 0 EUR
23016160.
Bench protectors, Benchkote® Plus, Whatman™
Surface Protectors Bench Protectors
Cytiva's Whatman™ Benchkote® Plus bench protector is a thicker, more absorbent version of the original Benchkote® bench paper. Designed for demanding or hazardous applications, it can absorb more than 750 ml/m² water.
  • Material is strong and tear resistant when wet or dry
  • Smooth white surface can be written on with ink or pencil and lies flat
  • Suitable for saturation with disinfectant to protect benches where pathogens and other bacteria are present
  • Can be incinerated after use: PE layer is rapidly consumed when incinerated and does not melt or drip