Dressing scissors
Supplier: BOCHEM
4103 4100 4101 4102 4104 4105 4106 4107 4108
231-1292EA 12 EUR
231-1292 231-1290 231-1291 BOCH4102 231-1293 BOCH4105 231-1294 231-1295 BOCH4108
Dressing scissors
Scissors
Stainless steel. Multi-purpose scissors for a wide range of applications.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Designed for rough handling
Order Now
SPECIFICATIONS
Environmentally Preferable