Disposable powder lances, SteriWare®
Supplier: Sampling Systems
8063H-1000 8063H-1000S 8063H-500 8063H-500S 8064H-500 8064H-1000S 8064H-500S 8063H-300 8064H-1000 8063H-300S
231-3500EA 384 EUR
231-3500 231-3501 231-3502 231-3503 SAMP8064H-500 SAMP8064H-1000S SAMP8064H-500S SAMP8063H-300 SAMP8064H-1000 SAMP8063H-300S
Disposable powder lances, SteriWare®
Samplers Disposable Samplers
These disposable powder lances are designed for taking cross sectional samples through a bulk powder.
  • Disposable items – No cleaning is required before or after sampling
  • Assembled and packed in a medical cleanroom
  • BSE/TSE free, full batch traceability
Suitable for industries such as pharmaceutical, food, chemical, cosmetic and biotechnology.
Certifications: FDA, EU 10/2011 and EC 1935/2004. ATEX Zones 0, 1, 2.
Order Now
SPECIFICATIONS
Environmentally Preferable