Dichloromethane ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals
Warning


Stabilization: 20 ppm Stabilised with 2-methyl-2-butene (iso-amylene)
Filtered through a 0.2 µm filter, packaged under nitrogen.

23373.320EA 106 EUR
23373.320 23373.290 23373.400
Dichloromethane ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Dichloromethane
Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
REACH: 01-2119480404-41
Merck Index: 12,06140
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=182687c44554cb33e52776bdf02ea799c8f367b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.8 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Evaporation residue Max. 10 ppm
Water Max. 0.05 %
Absorbance (240 nm) Max. 0.3
Absorbance (245 nm) Max. 0.1
Absorbance (260 nm) Max. 0.01
Transmittance (240 nm) Min. 50 %
Transmittance (245 nm) Min. 80 %
Transmittance (260 nm) Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR