Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate

Supplier: ALFA AESAR
Warning

24127.03
24127.03EA 0 EUR
24127.03
Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate
Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate

CAS No.: 16165-32-5 Molecular Formula: C6H24Cl3CrN6.3.5H2O Formula Weight: 401.71 (338.65anhy) Physical Form: Crystalline Appearance: Yellow to orange MDL No.: MFCD00150160

Formula: C₆H₂₄Cl₃CrN₆.3,5H₂O
MW: 401.71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150160
CAS Number: 16165-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR