1-Propanol ≥99% (by GC), Purum

Supplier: HONEYWELL
Danger

24135-160KG 24135-1L 24135-2.5L 24135-20KG 24135-6X1L
24135-160KGEA 2060 EUR
24135-160KG 24135-1L 24135-2.5L 24135-20KG 24135-6X1L
1-Propanol ≥99% (by GC), Purum
1-Propanol

  • ≥95% for general laboratory use.


Beilstein Number: 1098242
RTECS Number: UH8225000
UNSPSC Code: 12191601

Formula: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Boiling Pt: 96…98 °C (1.013 hPa)
Melting Pt: –127 °C
Density: 0.803…0.804 g/cm³ (20 °C)
Flash Pt: 21 °C
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3 II
Merck Index: 13,07934
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=b4a45c3a7a92d9e6aed8d1d3bcf6b139b7a0e31f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99 %
Non-volatile matter Max. 0.005 %
Water (Karl Fischer) Max. 0.1 %
Free acid (as C₂H₅COOH) Max. 0.002 %
Autoignition Temperature 700 °F
Vapor Pressure 10 mmHg ( 147 °C)
Vapor Density 2.1 (vs air)
Refractive Index n20/D 1.384 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR