Pipette tray for drawer
Supplier: KARTELL
252
252.EA 111 EUR
252.
Pipette tray for drawer
Trays/Dishes/Pots Pipette Trays
PVC, white.
  • Designed to hold about 30 pipettes
  • Four compartments to accommodate 1, 2, 5, 10 ml pipettes
W×D×H: 425×300×30 mm.